Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ORP3C
Entrez Gene ID
836061
Gene Name
OSBP(oxysterol binding protein)-related protein 3C
Gene Aliases
AT5G59420, F2O15.17, F2O15_17, OSBP(oxysterol binding protein)-related protein 3C
Chromosome Location
Chr.5: 23961567 - 23964874 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125333.5 NP_200750.1 2 - 3 354 97
GenBank mRNA AY054473.1 - 2 - 3 307 97
BT000115.1 - 2 - 3 190 97

View Full Product Details