Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VIP2
Entrez Gene ID
836092
Gene Name
VIRE2 interacting protein 2
Gene Aliases
AT5G59710, AtVIP2, MTH12.8, MTH12_8, NOT2b, Negative on TATA less2b, VIRE2 interacting protein 2
Chromosome Location
Chr.5: 24057378 - 24062082 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001345367.1 NP_001329954.1 12 - 13 1924 77
NM_125363.3 NP_568912.2 11 - 12 2424 77

View Full Product Details