Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TET4
Entrez Gene ID
836144
Gene Name
tetraspanin4
Gene Aliases
AT5G60220, F15L12.11, F15L12_11, tetraspanin4
Chromosome Location
Chr.5: 24248920 - 24250117 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125415.2 NP_200830.1 1 - 2 552 98

View Full Product Details