Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ALP
Entrez Gene ID
836158
Gene Name
aleurain-like protease
Gene Aliases
AALP, AT5G60360, MUF9.4, MUF9_4, SAG2, SENESCENCE ASSOCIATED GENE2, aleurain-like protease
Chromosome Location
Chr.5: 24279660 - 24282482 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125429.4 NP_568921.1 7 - 8 1421 93
GenBank mRNA AF083703.1 - 7 - 8 1102 93
AF233883.1 - 7 - 8 1046 93
AF360273.1 - 7 - 8 1076 93
AK317029.1 - 7 - 8 1109 93
AK317496.1 - 7 - 8 1109 93
AY088662.1 - 7 - 8 1116 93
BT000673.1 - 7 - 8 1109 93
BT000676.1 - 7 - 8 1105 93

View Full Product Details