Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
bZIP70
Entrez Gene ID
836204
Gene Name
basic leucine-zipper 70
Gene Aliases
AT5G60830, AtbZIP70, MAE1.7, MAE1_7, basic leucine-zipper 70
Chromosome Location
Chr.5: 24472639 - 24473319 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125476.1 NP_200891.1 1 - 2 533 64

View Full Product Details