Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PDCB1
Entrez Gene ID
836234
Gene Name
plasmodesmata callose-binding protein 1
Gene Aliases
AT5G61130, AtPDCB1, MAF19.13, MAF19_13, plasmodesmata callose-binding protein 1
Chromosome Location
Chr.5: 24587173 - 24589580 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125506.5 NP_200921.2 2 - 3 498 61
GenBank mRNA BT003876.1 - 2 - 3 498 61
BT005195.1 - 2 - 3 287 61

View Full Product Details