Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MIM
- Entrez Gene ID
-
836267
- Gene Name
- P-loop containing nucleoside triphosphate hydrolases superfamily protein
- Gene Aliases
-
AT5G61460, ATRAD18, MCI2.2, MCI2_2, SMC6B, STRUCTURAL MAINTENANCE OF CHROMOSOMES 6B, hypersensitive to MMS, irradiation and MMC
- Chromosome Location
-
Chr.5: 24714076 - 24721970 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_125539.2 |
NP_200954.1 |
18 - 19 |
2419 |
95 |
View Full Product Details