Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABCA9
Entrez Gene ID
836295
Gene Name
ABC2 homolog 11
Gene Aliases
ABC2 homolog 11, ARABIDOPSIS THALIANA ABC2 HOMOLOG 11, AT5G61730, ATATH11, ATH11, ATP-binding cassette A9, AtABCA9, MAC9.4, MAC9_4
Chromosome Location
Chr.5: 24803427 - 24807920 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001345493.1 NP_001330984.1 11 - 12 1954 74
NM_125567.2 NP_200981.1 11 - 12 1902 74

View Full Product Details