Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABCA10
Entrez Gene ID
836296
Gene Name
ABC2 homolog 14
Gene Aliases
ABC2 homolog 14, ARABIDOPSIS THALIANA ABC2 HOMOLOG 14, AT5G61740, ATATH14, ATH14, ATP-binding cassette A10, MAC9.5, MAC9_5
Chromosome Location
Chr.5: 24808406 - 24812739 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001345494.1 NP_001318860.1 5 - 6 812 97
NM_001345495.1 NP_001332100.1 5 - 6 632 97
NM_001345496.1 NP_001332101.1 3 - 4 356 97

View Full Product Details