Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LFY
Entrez Gene ID
836307
Gene Name
floral meristem identity control protein LEAFY (LFY)
Gene Aliases
AT5G61850, LEAFY, LEAFY 3, LFY3, MAC9.13, MAC9_13
Chromosome Location
Chr.5: 24844248 - 24846989 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125579.1 NP_200993.1 1 - 2 458 64
GenBank mRNA DQ447103.1 - 1 - 2 458 64
KF051022.1 - 1 - 2 458 64

View Full Product Details