Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AGD1
Entrez Gene ID
836319
Gene Name
ARF-GAP domain 1
Gene Aliases
ARF-GAP domain 1, AT5G61980, K22G18.9, K22G18_9
Chromosome Location
Chr.5: 24894053 - 24899973 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125591.3 NP_201004.2 6 - 7 817 81

View Full Product Details