Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAP65-9
Entrez Gene ID
836346
Gene Name
microtubule-associated protein 65-9
Gene Aliases
AT5G62250, MMI9.8, MMI9_8, microtubule-associated protein 65-9
Chromosome Location
Chr.5: 25005434 - 25008361 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001345543.1 NP_001318863.1 8 - 9 1486 143
NM_125619.2 NP_201031.1 7 - 8 1509 143
GenBank mRNA DQ056733.1 - 7 - 8 1379 143

View Full Product Details