Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- IRE
- Entrez Gene ID
-
836352
- Gene Name
- AGC (cAMP-dependent, cGMP-dependent and protein kinase C) kinase family protein
- Gene Aliases
-
AT5G62310, INCOMPLETE ROOT HAIR ELONGATION, MMI9.15, MMI9_15
- Chromosome Location
-
Chr.5: 25023155 - 25028686 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_125625.3 |
NP_201037.1 |
8 - 9 |
2834 |
117 |
View Full Product Details