Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LACS2
Entrez Gene ID
841367
Gene Name
long-chain acyl-CoA synthetase 2
Gene Aliases
AT1G49430, F13F21.14, F13F21_14, LATERAL ROOT DEVELOPMENT 2, LRD2, long-chain acyl-CoA synthetase 2
Chromosome Location
Chr.1: 18290942 - 18295905 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001333383.1 NP_001321632.1 11 - 12 1202 159
NM_103833.5 NP_175368.2 11 - 12 1239 159
GenBank mRNA AF503752.1 - 11 - 12 993 159
AY065424.1 - 11 - 12 1137 159
AY094420.1 - 11 - 12 1074 159
BT001970.1 - 11 - 12 993 159

View Full Product Details