Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LACS2
Entrez Gene ID
841367
Gene Name
long-chain acyl-CoA synthetase 2
Gene Aliases
AT1G49430, F13F21.14, F13F21_14, LATERAL ROOT DEVELOPMENT 2, LRD2, long-chain acyl-CoA synthetase 2
Chromosome Location
Chr.1: 18290942 - 18295905 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001333383.1 NP_001321632.1 2 - 3 423 67
NM_103833.5 NP_175368.2 2 - 3 460 67
GenBank mRNA AF503752.1 - 2 - 3 214 67
AY065424.1 - 2 - 3 358 67
AY094420.1 - 2 - 3 295 67
BT001970.1 - 2 - 3 214 67

View Full Product Details