Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NQR
Entrez Gene ID
841391
Gene Name
ARP protein (REF)
Gene Aliases
AT1G49670, F14J22.10, F14J22_10
Chromosome Location
Chr.1: 18380804 - 18386146 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198253.1 NP_001185182.1 5 - 6 676 146
NM_103855.5 NP_175390.2 5 - 6 714 146
GenBank mRNA AY062728.1 - 5 - 6 670 146
AY072306.1 - 5 - 6 675 146
AY093342.1 - 5 - 6 589 146
BT008393.1 - 5 - 6 589 146

View Full Product Details