Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NQR
Entrez Gene ID
841391
Gene Name
ARP protein (REF)
Gene Aliases
AT1G49670, F14J22.10, F14J22_10
Chromosome Location
Chr.1: 18380804 - 18386146 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198253.1 NP_001185182.1 7 - 8 947 111
NM_103855.5 NP_175390.2 7 - 8 985 111
GenBank mRNA AY062728.1 - 7 - 8 941 111
AY072306.1 - 7 - 8 946 111
AY093342.1 - 7 - 8 860 111
BT008393.1 - 7 - 8 860 111

View Full Product Details