Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SIRB
Entrez Gene ID
841439
Gene Name
sirohydrochlorin ferrochelatase B
Gene Aliases
AT1G50170, ATSIRB, F14I3.20, F14I3_20, sirohydrochlorin ferrochelatase B
Chromosome Location
Chr.1: 18581998 - 18583832 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001333424.1 NP_001323380.1 1 - 2 138 96
NM_103902.4 NP_564562.1 1 - 2 148 96
GenBank mRNA BT004060.1 - 1 - 2 75 96
BT005145.1 - 1 - 2 59 96

View Full Product Details