Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
THX
Entrez Gene ID
841454
Gene Name
thioredoxin X
Gene Aliases
AT1G50320, ATHX, ATX, THIOREDOXIN X, thioredoxin X
Chromosome Location
Chr.1: 18638384 - 18639536 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_103916.3 NP_564566.1 1 - 2 410 85
GenBank mRNA AF324698.2 - 1 - 2 384 85
AF326860.1 - 1 - 2 384 85
AF339683.1 - 1 - 2 338 85
AF386924.1 - 1 - 2 385 85
BT000355.1 - 1 - 2 338 85

View Full Product Details