Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- HAM2
- Entrez Gene ID
-
830834
- Gene Name
- histone acetyltransferase of the MYST family 2
- Gene Aliases
-
AT5G09740, F17I14.70, F17I14_70, HAC11, HAG05, HAG5, HISTONE ACETYLTRANSFERASE OF THE CBP FAMILY 11, HISTONE ACETYLTRANSFERASE OF THE GNAT/MYST SUPERFAMILY 5, histone acetyltransferase of the MYST family 2
- Chromosome Location
-
Chr.5: 3021932 - 3024763 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_121011.4 |
NP_196536.1 |
5 - 6 |
783 |
99 |
GenBank mRNA |
AB493744.1 |
- |
5 - 6 |
731 |
99 |
View Full Product Details