Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SULTR2;1
Entrez Gene ID
830882
Gene Name
slufate transporter 2;1
Gene Aliases
ARABIDOPSIS SULFATE TRANSPORTER 68, AST68, AT5G10180, slufate transporter 2;1, sulfate transporter 2;1
Chromosome Location
Chr.5: 3193017 - 3197136 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_121056.3 NP_196580.1 2 - 3 657 102
GenBank mRNA AB003591.1 - 2 - 3 516 102
AY062546.1 - 2 - 3 530 102
AY093335.1 - 2 - 3 449 102

View Full Product Details