Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RABH1e
Entrez Gene ID
830890
Gene Name
RAB GTPase homolog H1E
Gene Aliases
AT5G10260, AtRABH1e, F18D22.30, F18D22_30, RAB GTPase homolog H1E
Chromosome Location
Chr.5: 3219677 - 3221515 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_121064.4 NP_196588.3 2 - 3 503 140

View Full Product Details