Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CDKC;1
Entrez Gene ID
830891
Gene Name
cyclin-dependent kinase C;1
Gene Aliases
AT5G10270, F18D22.40, F18D22_40, cyclin-dependent kinase C;1
Chromosome Location
Chr.5: 3221493 - 3224788 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_121065.3 NP_196589.1 9 - 10 989 83
GenBank mRNA AF375437.1 - 9 - 10 966 83
AY120690.1 - 9 - 10 875 83

View Full Product Details