Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HK5
Entrez Gene ID
830938
Gene Name
histidine kinase 5
Gene Aliases
AHK5, AT5G10720, CKI2, CYTOKININ INDEPENDENT 2, MAJ23.80, MAJ23_80, histidine kinase 5
Chromosome Location
Chr.5: 3385901 - 3390686 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_121110.2 NP_196633.2 3 - 4 1137 106
GenBank mRNA DQ167579.1 - 3 - 4 1137 106

View Full Product Details