Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ER
Entrez Gene ID
830948
Gene Name
enhancer of rudimentary protein
Gene Aliases
ARABIDOPSIS THALIANA ENHANCER OF RUDIMENTARY HOMOLOGUE, AT5G10810, ATER, ENHANCER OF RUDIMENTARY HOMOLOG ATER, ENHANCER OF RUDIMENTARY HOMOLOGUE, T30N20.80, T30N20_80
Chromosome Location
Chr.5: 3418610 - 3420692 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_121119.3 NP_568240.1 2 - 3 249 134
GenBank mRNA AK117573.1 - 2 - 3 192 134
BT005033.1 - 1 - 2 113 134
U67398.1 - 2 - 3 151 134

View Full Product Details