Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SYTD
- Entrez Gene ID
-
830978
- Gene Name
- Calcium-dependent lipid-binding (CaLB domain) family protein
- Gene Aliases
-
AT5G11100, ATSYTD, NTMC2T2.2, NTMC2TYPE2.2, SYT4, T5K6.90, T5K6_90, synaptotagmin 4
- Chromosome Location
-
Chr.5: 3532402 - 3535221 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_121148.3 |
NP_196671.2 |
4 - 5 |
381 |
144 |
GenBank mRNA |
AM410051.1 |
- |
4 - 5 |
382 |
144 |
View Full Product Details