Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SPS2F
- Entrez Gene ID
-
830979
- Gene Name
- sucrose phosphate synthase 2F
- Gene Aliases
-
AT5G11110, ATSPS2F, KAONASHI 2, KNS2, SPS1, SPSA2, SUCROSE PHOSPHATE SYNTHASE 1, T5K6.100, T5K6_100, sucrose phosphate synthase 2F, sucrose-phosphate synthase A2
- Chromosome Location
-
Chr.5: 3536189 - 3541149 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_121149.4 |
NP_196672.3 |
12 - 13 |
3098 |
60 |
GenBank mRNA |
KJ138698.1 |
- |
12 - 13 |
2861 |
60 |
View Full Product Details