Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
APT5
Entrez Gene ID
830985
Gene Name
adenine phosphoribosyltransferase 5
Gene Aliases
AT5G11160, F2I11.50, F2I11_50, adenine phosphoribosyltransferase 5
Chromosome Location
Chr.5: 3550300 - 3552255 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001343159.1 NP_001329978.1 2 - 3 673 109
NM_121154.4 NP_196677.1 2 - 3 673 109
GenBank mRNA BT004028.1 - 2 - 3 423 109
BT005077.1 - 2 - 3 199 109

View Full Product Details