Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- DXPS3
- Entrez Gene ID
-
831009
- Gene Name
- 1-deoxy-D-xylulose 5-phosphate synthase 3
- Gene Aliases
-
1-deoxy-D-xylulose 5-phosphate (DXP) synthase 3, 1-deoxy-D-xylulose 5-phosphate synthase 3, AT5G11380, DXS3, F2I11.270, F2I11_270
- Chromosome Location
-
Chr.5: 3629988 - 3633511 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_121176.4 |
NP_196699.1 |
8 - 9 |
1857 |
68 |
View Full Product Details