Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DGS1
Entrez Gene ID
831104
Gene Name
dgd1 suppressor 1
Gene Aliases
AT5G12290, DGD1 SUPPRESSOR 1
Chromosome Location
Chr.5: 3974119 - 3978221 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001343232.1 NP_001330601.1 10 - 11 1511 83
NM_001343233.1 NP_001330602.1 10 - 11 1511 83
NM_001343234.1 NP_001330600.1 10 - 11 1511 83
NM_121267.5 NP_568262.2 10 - 11 1511 83
GenBank mRNA BT002450.1 - 10 - 11 1440 83
BT008365.1 - 10 - 11 1356 83

View Full Product Details