Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SEC10
Entrez Gene ID
831112
Gene Name
exocyst complex component sec10
Gene Aliases
AT5G12370, exocyst complex component sec10
Chromosome Location
Chr.5: 4002742 - 4008804 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036794.2 NP_001031871.1 10 - 11 1270 66
NM_001036795.2 NP_001031872.1 10 - 11 1141 66
NM_121275.4 NP_568270.3 10 - 11 1142 66
GenBank mRNA AK222187.1 - 10 - 11 1247 66

View Full Product Details