Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HA5
Entrez Gene ID
816988
Gene Name
plasma membrane H+-ATPase
Gene Aliases
AHA5, AT2G24520, F25P17.18, F25P17_18, H(+)-ATPase 5
Chromosome Location
Chr.2: 10414963 - 10420029 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001335933.1 NP_001318282.1 7 - 8 1795 82
NM_001335934.1 NP_001325336.1 8 - 9 1825 82
NM_001335935.1 NP_001325337.1 4 - 5 1326 82
NM_128013.2 NP_180028.1 6 - 7 1726 82

View Full Product Details