Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GPX1
Entrez Gene ID
817046
Gene Name
glutathione peroxidase 1
Gene Aliases
AT2G25080, ATGPX1, F13D4.40, F13D4_40, GLUTATHIONE PEROXIDASE 1, glutathione peroxidase 1
Chromosome Location
Chr.2: 10667898 - 10670127 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_128065.5 NP_180080.1 5 - 6 920 98
GenBank mRNA AY035153.1 - 5 - 6 733 98

View Full Product Details