Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- RLP20
- Entrez Gene ID
-
817082
- Gene Name
- receptor like protein 20
- Gene Aliases
-
AT2G25440, AtRLP20, F13B15.10, F13B15_10, receptor like protein 20
- Chromosome Location
-
Chr.2: 10826735 - 10829402 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_128101.1 |
NP_180114.1 |
2 - 3 |
559 |
143 |
View Full Product Details