Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IQD6
Entrez Gene ID
817158
Gene Name
IQ-domain 6
Gene Aliases
AT2G26180, IQ-domain 6, T1D16.18, T1D16_18
Chromosome Location
Chr.2: 11143290 - 11145103 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_128176.3 NP_180187.1 1 - 2 190 84
GenBank mRNA BT029734.1 - 1 - 2 69 84

View Full Product Details