Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARSK1
Entrez Gene ID
817169
Gene Name
root-specific kinase 1
Gene Aliases
AT2G26290, T1D16.7, T1D16_7, root-specific kinase 1
Chromosome Location
Chr.2: 11191978 - 11194431 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_128186.3 NP_180197.1 3 - 4 858 104

View Full Product Details