Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPN13
Entrez Gene ID
817200
Gene Name
regulatory particle non-ATPase 13
Gene Aliases
AT2G26590, T9J22.26, T9J22_26, regulatory particle non-ATPase 13
Chromosome Location
Chr.2: 11310957 - 11315524 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036347.2 NP_001031424.1 4 - 5 565 68
NM_001036348.2 NP_001031425.1 5 - 6 861 68
NM_001336077.1 NP_001325357.1 5 - 6 398 68
NM_128216.3 NP_565626.1 5 - 6 826 68

View Full Product Details