Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- GRV2
- Entrez Gene ID
-
817230
- Gene Name
- DNAJ heat shock N-terminal domain-containing protein
- Gene Aliases
-
AT2G26890, F12C20.7, F12C20_7, GRAVITROPISM DEFECTIVE 2, KAM2, KATAMARI2
- Chromosome Location
-
Chr.2: 11462067 - 11474371 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_128246.5 |
NP_180257.3 |
14 - 15 |
6120 |
92 |
View Full Product Details