The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_128277.5 | NP_180287.2 | 2 - 3 | 677 | 94 |
GenBank mRNA | AY065067.1 | - | 2 - 3 | 513 | 94 |
AY133599.1 | - | 2 - 3 | 497 | 94 |