Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PAP12
Entrez Gene ID
817261
Gene Name
purple acid phosphatase 12
Gene Aliases
ARABIDOPSIS THALIANA PURPLE ACID PHOSPHATASE 1, AT2G27190, ATPAP1, ATPAP12, PAP1, PURPLE ACID PHOSPHATASE 1, PURPLE ACID PHOSPHATASE 12, SECRETED PURPLE ACID PHOSPHATASE PRECURSOR, T22O13.4, T22O13_4, purple acid phosphatase 12
Chromosome Location
Chr.2: 11621222 - 11623618 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_128277.5 NP_180287.2 5 - 6 1268 83
GenBank mRNA AY065067.1 - 5 - 6 1104 83
AY133599.1 - 5 - 6 1088 83

View Full Product Details