Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CLV3
Entrez Gene ID
817267
Gene Name
CLAVATA3
Gene Aliases
AT2G27250, AtCLV3, CLAVATA3, F12K2.17, F12K2_17
Chromosome Location
Chr.2: 11664978 - 11665797 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_128283.2 NP_565641.2 1 - 1 341 142

View Full Product Details