Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AXS1
Entrez Gene ID
817332
Gene Name
UDP-D-apiose/UDP-D-xylose synthase 1
Gene Aliases
AT2G27860, F15K20.4, F15K20_4, UDP-D-apiose/UDP-D-xylose synthase 1
Chromosome Location
Chr.2: 11864277 - 11866998 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_128345.4 NP_180353.1 7 - 8 1076 78
GenBank mRNA AF361574.1 - 7 - 8 973 78
AY086830.1 - 7 - 8 979 78
AY442191.1 - 7 - 8 921 78
AY735589.1 - 7 - 8 985 78
AY924751.1 - 7 - 8 921 78
BT001016.1 - 7 - 8 921 78

View Full Product Details