Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SCPL51
Entrez Gene ID
817336
Gene Name
serine carboxypeptidase-like 51
Gene Aliases
AT2G27920, T1E2.16, T1E2_16, serine carboxypeptidase-like 51
Chromosome Location
Chr.2: 11885571 - 11889176 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036357.1 NP_001031434.1 12 - 13 1298 153
NM_128351.3 NP_565663.1 12 - 13 1454 153
NM_201822.1 NP_973551.1 10 - 11 1119 153
GenBank mRNA AF412049.1 - 12 - 13 1391 153

View Full Product Details