Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SCPL51
Entrez Gene ID
817336
Gene Name
serine carboxypeptidase-like 51
Gene Aliases
AT2G27920, T1E2.16, T1E2_16, serine carboxypeptidase-like 51
Chromosome Location
Chr.2: 11885571 - 11889176 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036357.1 NP_001031434.1 2 - 3 215 83
NM_128351.3 NP_565663.1 2 - 3 361 83
GenBank mRNA AF412049.1 - 2 - 3 298 83
BT003041.1 - 2 - 3 228 83

View Full Product Details