Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATCHX8
Entrez Gene ID
817364
Gene Name
cation/hydrogen exchanger family protein
Gene Aliases
AT2G28180, CATION/H+ EXCHANGER 8, CATION/H+ EXCHANGER 8, CHX08, CHX8, F24D13.3, F24D13_3
Chromosome Location
Chr.2: 12010994 - 12013739 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_128378.4 NP_180385.3 1 - 2 301 144
GenBank mRNA AY926468.1 - 1 - 2 301 144

View Full Product Details