Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CNGC15
Entrez Gene ID
817372
Gene Name
cyclic nucleotide-gated channel 15
Gene Aliases
AT2G28260, ATCNGC15, T3B23.7, T3B23_7, cyclic nucleotide-gated channel 15
Chromosome Location
Chr.2: 12049443 - 12052643 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336157.1 NP_001324840.1 4 - 5 1550 128
NM_128386.2 NP_180393.1 5 - 6 1950 128

View Full Product Details