Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARF10
Entrez Gene ID
817382
Gene Name
auxin response factor 10
Gene Aliases
AT2G28350, T1B3.13, T1B3_13, auxin response factor 10
Chromosome Location
Chr.2: 12113889 - 12116957 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336164.1 NP_001324968.1 2 - 3 2201 99
NM_128394.5 NP_180402.1 2 - 3 2374 99
GenBank mRNA AB493569.1 - 2 - 3 1932 99
AF325073.1 - 2 - 3 1932 99
AF336919.1 - 2 - 3 1932 99

View Full Product Details