Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VLN1
Entrez Gene ID
817539
Gene Name
villin-like 1
Gene Aliases
AT2G29890, ATVLN1, F6K5.2, F6K5_2, villin 1, villin-like 1
Chromosome Location
Chr.2: 12744116 - 12749596 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036367.2 NP_001031444.2 13 - 14 1762 91
NM_001202706.1 NP_001189635.1 13 - 14 1762 91
NM_001336227.1 NP_001324174.1 13 - 14 1745 91
NM_001336228.1 NP_001324173.1 13 - 14 1847 91
NM_128543.4 NP_029567.1 13 - 14 1781 91

View Full Product Details