Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VLN1
Entrez Gene ID
817539
Gene Name
villin-like 1
Gene Aliases
AT2G29890, ATVLN1, F6K5.2, F6K5_2, villin 1, villin-like 1
Chromosome Location
Chr.2: 12744116 - 12749596 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036367.2 NP_001031444.2 9 - 10 1238 79
NM_001202706.1 NP_001189635.1 9 - 10 1238 79
NM_001336227.1 NP_001324174.1 9 - 10 1272 79
NM_001336228.1 NP_001324173.1 9 - 10 1323 79
NM_128543.4 NP_029567.1 9 - 10 1257 79

View Full Product Details