Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AHP1
Entrez Gene ID
821705
Gene Name
histidine-containing phosphotransmitter 1
Gene Aliases
AT3G21510, histidine-containing phosphotransmitter 1
Chromosome Location
Chr.3: 7577712 - 7579789 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_113046.4 NP_188788.1 1 - 2 312 140
GenBank mRNA AB012570.1 - 1 - 2 94 140
AB015141.1 - 1 - 2 100 140
AF370265.1 - 1 - 2 109 140
AY063082.1 - 1 - 2 60 140

View Full Product Details